Novus Biologicals
Manufacturer Code:NBP191229
Catalog # NBP191229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LLPEERLRNLFSYDGIWLFPKNQCKCEANKEQGGYNFQDAYGQSDLPAVKARRQAEFEHFQRREGLPRPLPLLVQPNLPFGYPVHGVEVMPLHTVPIPGLQFEGPDA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4 GalNAcT/Sda-GalNAcT B4GALT beta 14 N-acetylgalactosaminyltransferase/betal beta-14 N-acetylgalactosaminyltransferase 2 beta-14-N-acetyl-galactosaminyl transferase 2 beta-4-N-acetylgalactosaminyltransferase GALGT2 MGC142235 MGC142237 sd(a) beta-14-GalNAc transferase sda beta-14-GalNAc transferase UDP-GalNAc:Neu5Aca2-3Galb-R b14-N-acetylgalactosaminyltransferase; beta 1,4 N-acetylgalactosaminyltransferase/betal,4 GalNAcT/Sda-GalNAcT; Beta-1,4 N-acetylgalactosaminyltransferase 2; beta-1,4-N-acetyl-galactosaminyl transferase 2; beta-4-N-acetylgalactosaminyltransferase; Sd(a) beta-1,4-GalNAc transferase; sda beta-1,4-GalNAc transferase; UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase; UDP-GalNAc:Neu5Acalpha2-3Galbeta-R beta1,4-N-acetylgalactosaminyltransferase
Gene Aliases: B4GALNT2; B4GALT; GALGT2
UniProt ID: (Human) Q8NHY0
Entrez Gene ID: (Human) 124872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.