Novus Biologicals
Manufacturer Code:NBP297401H
Catalog # NB042927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human B4GALNT1 fragment. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (N-acetylneuraminyl)-galactosylglucosylceramide; (N-acetylneuraminyl)-galactosylglucosylceramide beta-14 N-acetylgalactosaminyltransferase 1 beta14GalNAc-T beta-14-N-acetyl-galactosaminyl transferase 1 beta1-4GalNAc-T GALGTEC 2.4.1.92 GalNAc-T GALNACT GD2 synthase GM2 synthase GM2/GD2 synthase SIAT2 UDP-Gal:betaGlcNAc beta-14-N-acetylgalactosaminyltransferase transferase 1 UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramideN-acetylgalactosaminyltransferase (GalNAc-T); Beta-1,4 N-acetylgalactosaminyltransferase 1; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta1,4GalNAc-T; GalNAc-T; GD2 synthase, GM2 synthase; GM2/GD2 synthase; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T)
Gene Aliases: B4GALNT1; GALGT; GalNAc-T; GALNACT; SIAT2; SPG26
UniProt ID: (Human) Q00973
Entrez Gene ID: (Human) 2583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.