Abnova
Manufacturer Code:H00002583A01
Catalog # 89-104-529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 30 a.a. to 127 a.a) partial recombinant protein with GST tag.,B4GALNT1 (NP_001469 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant B4GALNT1.
For Research Use Only
Protein Aliases: (N-acetylneuraminyl)-galactosylglucosylceramide; Beta-1,4 N-acetylgalactosaminyltransferase 1; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta1,4GalNAc-T; GALGT; GalNAc-T; GALNACT; GD2 synthase, GM2 synthase; GM2/GD2 synthase; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T)
Gene Aliases: B4GALNT1; GALGT; GalNAc-T; GALNACT; SIAT2; SPG26
UniProt ID: (Human) Q00973
Entrez Gene ID: (Human) 2583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.