Abnova
Manufacturer Code:MAB2415
Catalog # 89-110-996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | MC-631 |
Immunogen | Native purified from 4-8 cell stage mouse embryos. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against native B3galt5.
For Research Use Only
Protein Aliases: 1190002B21Rik; AU045265; b3Gal-T5; b3Galt-V; Beta-1,3-galactosyltransferase 5; Beta-1,3-GalTase 5; Beta-3-Gx-T5; beta3Gal-T5; beta3GalT5; SSEA-3 synthase; Stage-specific embryonic antigen 3 synthase; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 5; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 5
Gene Aliases: 1190002B21Rik; AU045265; b3Galt-V; B3galt5; B3gt5
UniProt ID: (Mouse) Q9JI67
Entrez Gene ID: (Mouse) 93961
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.