Invitrogen
Manufacturer Code:PA569352
Catalog # PIPA569352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat B3galnt1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b3Gal-T3; B3GALNT1 B3GALT3 UNQ531/PRO1074; Beta-1,3-galactosyltransferase 3; Beta-1,3-GalNAc-T1; Beta-1,3-GalTase 3; beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group); beta-3-GalNAc-T1; Beta-3-Gx-T3; beta3Gal-T3; beta3GalT3; Brainiac 1; brainiac1; Galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase; Globoside blood group; Globoside synthase; globotriaosylceramide 3-beta-N-acetylgalactosaminyltransferase; Mbrn 1; P antigen synthase; P blood group globoside; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase III; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 3; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 3 (Globoside blood group); UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 1; UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 1; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 1 (Globoside blood group); UDP-N-acetylgalactosamine:globotriaosylceramide beta-1,3-N-acetylgalactosaminyltransferase
Gene Aliases: B3GALNT1; B3GALT3; B3gt3; beta3Gal-T3; galT3; Gb4Cer; GLCT3; GLOB; Mbrn1; P; P1; UNQ531/PRO1074
UniProt ID: (Human) O75752, (Mouse) Q920V1, (Rat) Q6AY39
Entrez Gene ID: (Bovine) 767844, (Dog) 100683634, (Rabbit) 100352272, (Horse) 100062101, (Human) 8706, (Mouse) 26879, (Rat) 310508, (Guinea pig) 100729840
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.