Novus Biologicals
Manufacturer Code:NBP292163100UL
Catalog # NB026217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 36-378 of human B3GNT5 (NP_114436.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B3GN-T5 beta 13 N-acetyglucosaminyltransferase Lc3 synthase beta-13-Gn-T5 Beta-13-N-acetylglucosaminyltransferase 5 beta-13-N-acetylglucosaminyltransferase bGnT-5 beta3Gn-T5 BGnT-5 EC 2.4.1 EC 2.4.1.- Lactotriaosylceramide synthase Lc(3)Cer synthase Lc3 synthase UDP-GlcNAc:betaGal beta-13-N-acetylglucosaminyltransferase 5; beta 1,3 N-acetyglucosaminyltransferase Lc3 synthase; beta-1,3-Gn-T5; beta-1,3-N-acetylglucosaminyltransferase 5; beta-1,3-N-acetylglucosaminyltransferase bGnT-5; BGnT-5; Lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase; Lactotriaosylceramide synthase; Lc(3)Cer synthase; lc3 synthase; UDP-GlcNAc:beta-Gal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases: B3GN-T5; B3GNT5; beta3Gn-T5
UniProt ID: (Human) Q9BYG0
Entrez Gene ID: (Human) 84002
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.