Proteintech
Manufacturer Code:204221AP150UL
Catalog # 50-173-2881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | B3GNT5 Fusion Protein Ag14246 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B3GN T5 B3GNT5 Beta 13 Gn T5 beta3Gn T5 BGnT 5 Lactotriaosylceramide synthase Lc(3)Cer synthase Lc3 synthase; beta 1,3 N-acetyglucosaminyltransferase Lc3 synthase; beta-1,3-Gn-T5; beta-1,3-N-acetylglucosaminyltransferase 5; beta-1,3-N-acetylglucosaminyltransferase bGnT-5; beta-1,3-N-acetylglucosaminyltransferase-5; beta3Gn-T5; BGnT-5; Lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase; Lactotriaosylceramide synthase; Lc(3)Cer synthase; lc3 synthase; UDP-GlcNAc:beta-Gal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases: B3GN-T5; B3GNT5; beta3Gn-T5; beta3GnT5
UniProt ID: (Human) Q9BYG0, (Mouse) Q8BGY6, (Rat) Q99NB2
Entrez Gene ID: (Human) 84002, (Mouse) 108105, (Rat) 116740
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.