Invitrogen
Manufacturer Code:PA524189
Catalog # PIPA524189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 233-260 amino acids from the central region of human B3GNT4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B3GN-T4 beta3Gn-T4 BGnT-4 beta-13-Gn-T4 beta-13-N-acetylglucosaminyltransferase 4 beta-13-N-acetylglucosaminyltransferase bGn-T4; beta-1,3-Gn-T4; beta-1,3-N-acetylglucosaminyltransferase 4; beta-1,3-N-acetylglucosaminyltransferase bGn-T4; beta3Gn-T4; BGnT-4; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 4; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases: 1010001G17Rik; B3GN-T4; B3GNT4; beta3Gn-T4; BGnT-4; UNQ1898/PRO4344
UniProt ID: (Human) Q9C0J1, (Mouse) Q1RLK6
Entrez Gene ID: (Human) 79369, (Mouse) 231727
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.