Novus Biologicals
Manufacturer Code:NBP299178H
Catalog # NB058710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human B3GNT3. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B3GALT8 B3GAL-T8 B3GN-T3 B3GNT-3 Beta-13-galactosyltransferase 8 Beta-13-GalTase 8 Beta-13-Gn-T3 Beta-13-N-acetylglucosaminyltransferase 3 beta-13-N-acetylglucosaminyltransferase bGnT-3 Beta3GalT8 Beta3Gal-T8 beta3Gn-T3 Beta-3-Gx-T8 BGnT-3 Core 1 extending beta-13-N-acetylglucosaminyltransferase core1-beta3GlcNAcT EC 2.4.1 EC 2.4.1.- HP10328 putative type II membrane protein TMEM3beta3Gal-T8 transmembrane protein 3 UDP-Gal:beta-GlcNAc beta-13-galactosyltransferase 8 UDP-galactose:beta-N-acetylglucosamine beta-13-galactosyltransferase 8 UDP-GlcNAc:betaGal beta-13-N-acetylglucosaminyltransferase 3; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; Beta-1,3-galactosyltransferase 8; Beta-1,3-GalTase 8; beta-1,3-Gn-T3; beta-1,3-N-acetylglucosaminyltransferase bGnT-3; Beta-3-Gx-T8; beta3GalT8; BGnT-3; Core 1 extending beta-1,3-N-acetylglucosaminyltransferase; Core1-beta3GlcNAcT; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 3; putative type II membrane protein; Transmembrane protein 3; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Gene Aliases: B3GAL-T8; B3GALT8; B3GN-T3; B3GNT-3; B3GNT3; beta3Gn-T3; HP10328; TMEM3; UNQ637/PRO1266
UniProt ID: (Human) Q9Y2A9
Entrez Gene ID: (Human) 10331
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.