OriGene
Manufacturer Code:TA505270
Catalog # 50-167-5787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G10 |
Immunogen | Full length human recombit protein of human B3GNT2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-Gn-T1 3-Gn-T2 B3GN-T2 B3GNT B3GNT-2 B3GNT1 BETA3GNT BGNT2 BGnT-2 beta-1 beta3Gn-T1 beta3Gn-T2; Beta-1,3-galactosyltransferase 7; Beta-1,3-GalTase 7; beta-1,3-Gn-T1; beta-1,3-Gn-T2; Beta-1,3-N-acetylglucosaminyltransferase 1; beta-1,3-N-acetylglucosaminyltransferase bGnT-1; beta-1,3-N-acetylglucosaminyltransferase bGnT-2; Beta-3-Gx-T7; BGnT-1; BGnT-2; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 2; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2
Gene Aliases: 3-Gn-T1; 3-Gn-T2; B3GALT7; B3GN-T2; B3GNT; B3GNT-2; B3GNT1; B3GNT2; beta-1; beta3Gn-T1; beta3Gn-T2; BETA3GNT; BGnT-2; BGNT2
UniProt ID: (Human) Q9NY97
Entrez Gene ID: (Human) 10678
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.