Invitrogen
Manufacturer Code:PA542943
Catalog # PIPA542943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human B3GAT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-beta-glucuronosyltransferase; Beta-1 3-glucuronyltransferase 3 (glucuronosyltransferase I) GLCATI GlcAT-I; Beta-1,3-glucuronyltransferase 3; beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I); Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; GlcAT-I; GlcUAT-I; Glucuronosyltransferase I; glucuronosyltransferase-I; glucuronyltransferase I; Sqv-8-like protein; UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase
Gene Aliases: 2810405M13Rik; B3GAT3; GLCATI; glcUAT-I; JDSCD
UniProt ID: (Human) O94766, (Mouse) P58158
Entrez Gene ID: (Goat) 102172516, (Bovine) 404162, (Horse) 100063587, (Human) 26229, (Rabbit) 100350294, (Dog) 483785, (Pig) 397642, (Guinea pig) 100731926, (Mouse) 72727, (Rat) 293722
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.