Invitrogen
Manufacturer Code:PA523973
Catalog # PIPA523973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 123-150 amino acids from the central region of human B3GAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-glucuronyltransferase 2; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 2; GlcAT-D; GlcAT-S; GLCATS UDP-glucuronosyltransferase S UDP-glucuronyltransferase S galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 2 glcAT-D glucuronosyltransferase S uridine diphosphate glucuronic acid:acceptor glucuronosyltransferase; glucuronosyltransferase S; glucuronosyltransferase-S; UDP-glucuronosyltransferase S; UDP-glucuronosyltransferase-S; UDP-glucuronyltransferase S; uridine diphosphate glucuronic acid:acceptor glucuronosyltransferase
Gene Aliases: B3GAT2; GlcAT-S; GLCATS; KIAA1963
UniProt ID: (Human) Q9NPZ5, (Mouse) P59270
Entrez Gene ID: (Human) 135152, (Mouse) 280645
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.