Invitrogen
Manufacturer Code:PA569377
Catalog # PIPA569377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse B3GALT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-galactosyltransferase 2; Beta-1,3-GalTase 2; beta-3-galt2; beta3Gal-T2; beta3GalT2; BETA3GALT2 GLCT2 beta3Gal-T2 beta-13-galactosyltransferase 2 UDP-Gal:betaGlcNAc beta 13-galactosyltransferase polypeptide 2 UDP-galactose:2-acetamido-2-deoxy-D-glucose 3beta-galactosyltransferase 2 beta-13-GalTase 2 beta-3-galt2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase-II; UDP-galactose:2-acetamido-2-deoxy-D-glucose 3beta-galactosyltransferase 2
Gene Aliases: B3GALT2; beta3Gal-T2; BETA3GALT2; GLCT2; zgc:76904
UniProt ID: (Human) O43825, (Mouse) O54905
Entrez Gene ID: (Bovine) 539815, (Rabbit) 100348508, (Horse) 100051213, (Sheep) 101121025, (Goat) 102190625, (Dog) 607356, (Human) 8707, (Mouse) 26878, (Guinea pig) 100713957, (Rat) 686081, (Zebrafish) 404633
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.