Proteintech
Manufacturer Code:171421AP150UL
Catalog # 50-173-1401
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | B3GALNT2 Fusion Protein Ag10907 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B3GalNAc T2 B3GALNT2 Beta 13 GalNAc T2; Beta-1,3-GalNAc-T2; Beta-1,3-N-acetylgalactosaminyltransferase II; UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 2; UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 2
Gene Aliases: A930105D20Rik; B3GalNAc-T2; B3GALNT2; C80633; D230016N13Rik; MDDGA11; RGD1306946
UniProt ID: (Human) Q8NCR0, (Mouse) Q8BG28
Entrez Gene ID: (Human) 148789, (Rat) 291212, (Mouse) 97884
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.