Invitrogen
Manufacturer Code:PA5109438
Catalog # PIPA5109438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 44-331 of human B3GALNT1 (NP_003772.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b3Gal-T3; B3galnt1 B3galt3 B3gt3 Beta-13-galactosyltransferase 3 Beta-13-GalNAc-T1 Beta-13-GalTase 3 beta3Gal-T3 Beta-3-Gx-T3 BOS_1144 Brainiac 1 Galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase galT3 Gb4Cer GLCT3 GLOB Globoside synthase Mbrn1 P P1 UDP-GalNAc:beta-13-N-acetylgalactosaminyltransferase 1 UDP-N-acetylgalactosamine:globotriaosylceramide beta-13-N-acetylgalactosaminyltransferase UNQ531/PRO1074; Beta-1,3-galactosyltransferase 3; Beta-1,3-GalNAc-T1; Beta-1,3-GalTase 3; Beta-3-Gx-T3; beta3GalT3; brainiac1; Galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase; Globoside synthase; globotriaosylceramide 3-beta-N-acetylgalactosaminyltransferase; P antigen synthase; P blood group globoside; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 3 (Globoside blood group); UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 1; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 1 (Globoside blood group); UDP-N-acetylgalactosamine:globotriaosylceramide beta-1,3-N-acetylgalactosaminyltransferase
Gene Aliases: B3GALNT1; B3GALT3; beta3Gal-T3; galT3; Gb4Cer; GLCT3; GLOB; P; P1; UNQ531/PRO1074
UniProt ID: (Human) O75752
Entrez Gene ID: (Human) 8706
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.