Invitrogen
Manufacturer Code:PA577292
Catalog # PIPA577292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KEAS RTR*SG GPKGS K corresponding to amino acid residues 243-257 of rat BKRB1 with replacement of cysteine 250 (C250 ) with serine (*S) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B1 bradykinin receptor; B1R; BK-1 receptor; BKRB1; bradykinin B1 receptor; bradykinin receptor, beta 1; KB1; kinin B1; Kinin B1 receptor
Gene Aliases: B1BKR; B1R; Bdkrb; Bdkrb1; BKR; BKR1; BRADYB1
UniProt ID: (Rat) P97583, (Mouse) Q61125
Entrez Gene ID: (Rat) 81509, (Mouse) 12061
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.