Abnova
Manufacturer Code:MAB1726
Catalog # 89-110-597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HI-12 |
Immunogen | Native purified rat Azin1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native Azin1.
For Research Use Only
Protein Aliases: antizyme inhibitor 1; AZI; Oazi; Oazin; ornithine decarboxylase antizyme inhibitor
Gene Aliases: Oazi; Oazin
Entrez Gene ID: (Rat) 58961
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.