R&D Systems
Manufacturer Code:IC6965G
Catalog # IC6965G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 713610 |
Immunogen | Phosphopeptide containing the human Axl Y779 site |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI323647 ARK AXL oncogene AXL receptor tyrosine kinase AXL transforming sequence/gene EC 2.7.10 EC 2.7.10.1 JTK11 Tyro7 tyrosine-protein kinase receptor UFO UFO UFOoncogene AXL; AXL oncogene; AXL transforming sequence/gene; Tyrosine-protein kinase receptor UFO
Gene Aliases: ARK; AXL; JTK11; Tyro7; UFO
UniProt ID: (Human) P30530
Entrez Gene ID: (Human) 558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.