Invitrogen
Manufacturer Code:MA514790
Catalog # PIMA514790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | E.882.7 |
Immunogen | Recombit protein consisting of the N-terminal region of human Axin1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Axin-1; axin-1 axis inhibition protein 1 axis inhibitor 1 fused mouse homolog of hAxin; Axis inhibition protein 1; axis inhibitor 1; fused, mouse, homolog of; GSK-3beta interacting protein rAxin; hAxin; Protein Fused; protein phosphatase 1, regulatory subunit 49; rAxin
Gene Aliases: AI316800; AXIN; AXIN1; Fu; fused; Kb; Ki; kinky; knobbly; PPP1R49
UniProt ID: (Human) O15169, (Rat) O70239, (Mouse) O35625
Entrez Gene ID: (Human) 8312, (Rat) 79257, (Mouse) 12005
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.