R&D Systems
Manufacturer Code:MAB6078
Catalog # MAB6078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 354214 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Axin-2 Tyr550-Gln760 Accession # Q9Y2T1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Axil; Axil axin 2 axin-2 Axin2 Axin-like protein Axis inhibition protein 2 Conductin DKFZp781B0869 MGC10366 MGC126582; Axin-2; Axin-like protein; Axis inhibition protein 2; Conductin
Gene Aliases: AXIL; AXIN2; ODCRCS
UniProt ID: (Human) Q9Y2T1
Entrez Gene ID: (Human) 8313
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.