Novus Biologicals
Manufacturer Code:NBP176526
Catalog # NBP176526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human AXIN2. The immunogen is located within amino acids 780 - 830 of AXIN2. Amino Acid Squence: KEQLSKKGNYRYYFKKASDE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axil; Axil axin 2 axin-2 Axin-like protein Axis inhibition protein 2 Conductin DKFZp781B0869 MGC10366 MGC126582; Axin-2; Axin-like protein; Axis inhibition protein 2; Conductin
Gene Aliases: AXIL; AXIN2; ODCRCS
UniProt ID: (Human) Q9Y2T1
Entrez Gene ID: (Human) 8313
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.