Novus Biologicals
Manufacturer Code:NBP131013
Catalog # NBP131013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Axin 1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: axin 1 axin-1 AXINaxis inhibitor 1 Axis inhibition protein 1 fused mouse homolog of hAxin MGC52315; Axin-1; Axis inhibition protein 1; axis inhibitor 1; fused, mouse, homolog of; hAxin; protein phosphatase 1, regulatory subunit 49
Gene Aliases: AXIN; AXIN1; PPP1R49
UniProt ID: (Human) O15169
Entrez Gene ID: (Human) 8312
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.