R&D Systems
Manufacturer Code:MAB6184
Catalog # MAB6184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 609930 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse AIRE Ser476-Ser552 (predicted) Accession # Q9Z0E3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AIRE APECED APECED protein APECEDAPSI APS1 APSI Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein autoimmune regulator autoimmune regulator (APECED protein)10APS1AIRE1 autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy) PGA1; APECED protein; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; Autoimmune regulator
Gene Aliases: AIRE; AIRE1; APECED; APS1; APSI; PGA1
UniProt ID: (Human) O43918
Entrez Gene ID: (Human) 326
Molecular Function: ligase transcription cofactor transcription factor ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.