Novus Biologicals
Manufacturer Code:NBP271303UV
Catalog # NBP271303UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B7 |
Immunogen | Full-length protein expressed in 293T cell transfected with human Aurora C expression vector. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AIE2AIK3 ARK3Serine/threonine-protein kinase aurora-C AurC aurora kinase CAurora/IPL1/Eg2 protein 2 Aurora/IPL1-related kinase 3 aurora-C Aurora-related kinase 3 EC 2.7.11 EC 2.7.11.1 serine/threonine kinase 13 (aurora/IPL1-like) serine/threonine-protein kinase 13 STK13ARK-3; ARK-3; Aurora 3; Aurora kinase C; aurora-related kinase 3; Aurora/IPL1-related kinase 3; Aurora/IPL1/Eg2 protein 2; epididymis secretory protein Li 90; serine/threonine kinase 13 (aurora/IPL1-like); Serine/threonine-protein kinase 13; Serine/threonine-protein kinase aurora-C
Gene Aliases: AIE2; AIK3; AIRK3; ARK3; AurC; AURKC; aurora-C; HEL-S-90; SPGF5; STK13
UniProt ID: (Human) Q9UQB9
Entrez Gene ID: (Human) 6795
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.