Invitrogen
Manufacturer Code:PA513720
Catalog # PIPA513720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 115-145 amino acids from the central region of human Aurora-C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARK-3; Aurora 3; aurora B; Aurora kinase C; aurora-related kinase 3; Aurora/IPL1-related kinase 3; Aurora/IPL1/Eg2 protein 1; Aurora/IPL1/Eg2 protein 2; Aurora/Ipl1/Eg2 protein 2 Serine/threonine protein kinase 13 Aurora/Ipl1-related kinase 3 Aurora-C; epididymis secretory protein Li 90; serine/threonine kinase 13 (aurora/IPL-like); serine/threonine kinase 13 (aurora/IPL1-like); Serine/threonine-protein kinase 13; Serine/threonine-protein kinase aurora-C
Gene Aliases: AIE1; AIE2; AIK3; AIRK3; ARK-3; ARK3; AurC; AURKC; aurora-C; HEL-S-90; IAK3; SPGF5; STK13
UniProt ID: (Human) Q9UQB9, (Mouse) Q9JLC2
Entrez Gene ID: (Human) 6795, (Mouse) 20871
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.