Invitrogen
Manufacturer Code:PA5114014
Catalog # PIPA5114014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human AURKC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIE1 AIE2 AIK2 AIK3 AIM1 AIM-1 AIRK2 AIRK3 ARK2 ARK-2 ARK3 ARK-3 AurC AURKB AURKC Aurora 1 Aurora 3 aurora B Aurora kinase B Aurora kinase C aurora/Ipl1/Eg2 protein 1 Aurora/IPL1/Eg2 protein 2 Aurora/IPL1-related kinase 3 aurora-C aurora-related kinase 3 epididymis secretory protein Li 90 HEL-S-90 IAK3 serine/threonine kinase 13 (aurora/IPL1-like) serine/threonine kinase 13 (aurora/IPL-like) Serine/threonine-protein kinase 13 Serine/threonine-protein kinase aurora-C SPGF5 STK1 STK-1 STK12 STK13 STK5; ARK-3; Aurora 3; Aurora kinase C; aurora-related kinase 3; Aurora/IPL1-related kinase 3; Aurora/IPL1/Eg2 protein 2; epididymis secretory protein Li 90; serine/threonine kinase 13 (aurora/IPL1-like); Serine/threonine-protein kinase 13; Serine/threonine-protein kinase aurora-C
Gene Aliases: AIE2; AIK3; AIRK3; ARK3; AurC; AURKC; aurora-C; HEL-S-90; SPGF5; STK13
UniProt ID: (Human) Q9UQB9
Entrez Gene ID: (Human) 6795
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.