Novus Biologicals
Manufacturer Code:NBP254478AF594
Catalog # NP24478A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AURKB/1521 |
Immunogen | Recombinant human Aurora B protein fragment (aa89-251) (exact sequence is proprietary) |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AIK2EC 2.7.11.1 AIM1Aik2 AIM-1STK-1 ARK2ARK-2 AurB aurkb-sv2 Aurora- and IPL1-like midbody-associated protein 1 aurora kinase BSerine/threonine-protein kinase aurora-B aurora kinase B-Sv1 aurora kinase B-Sv2 Aurora/IPL1-related kinase 2 aurora-1 aurora-B Aurora-related kinase 2 EC 2.7.11 IPL1 serine/threonine kinase 12 serine/threonine-protein kinase 12 STK12aurkb-sv1 STK5; AIM-1; ARK-2; Aurora 1; Aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora- and IPL1-like midbody-associated protein 1; aurora-1; aurora-B; aurora-related kinase 2; Aurora/IPL1-related kinase 2; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase aurora-B; STK-1
Gene Aliases: AIK2; AIM-1; AIM1; AIRK2; ARK2; AurB; AURKB; aurkb-sv1; aurkb-sv2; IPL1; PPP1R48; STK1; STK12; STK5
UniProt ID: (Human) C7G533
Entrez Gene ID: (Human) 9212
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.