Invitrogen
Manufacturer Code:MA535292
Catalog # PIMA535292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0018 |
Immunogen | A synthesized peptide derived from human Aurora B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AIE2 AIK1 AIK2 AIK3 AIM1 AIM-1 AIRK2 AIRK3 AL022959 ARK2 ARK-2 ARK3 ARK-3 AurB Aurkb aurkb-sv1 aurkb-sv2 AURKC Aurora 1 Aurora 3 Aurora- and IPL1-like midbody-associated protein 1 Aurora kinase B Aurora kinase C Aurora/IPL1-related kinase 2 BOS_18918 EC 2.7.11.1 IPL1 NP_004208 PPP1R48 Serine/threonine-protein kinase 12 Serine/threonine-protein kinase 5 Serine/threonine-protein kinase aurora-B STK1 STK-1 STK12 STK13 STK5; AIM-1; ARK-2; Aurora 1; Aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora- and IPL1-like midbody-associated protein 1; aurora-1; aurora-B; aurora-related kinase 2; Aurora/IPL1-related kinase 2; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase aurora-B; STK-1
Gene Aliases: AIK2; AIM-1; AIM1; AIRK2; ARK-2; ARK2; AurB; AURKB; aurkb-sv1; aurkb-sv2; IPL1; PPP1R48; STK-1; STK1; STK12; STK5
UniProt ID: (Human) C7G533, (Rat) O55099
Entrez Gene ID: (Human) 9212, (Rat) 114592
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.