Invitrogen
Manufacturer Code:MA515804
Catalog # PIMA515804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G10 |
Immunogen | Purified recombit fragment of human AURKA expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARK-1; Aurora 2; Aurora kinase A; aurora-A; aurora-related kinase 1; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; Breast tumor-amplified kinase; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6 AIK ARK1 AURA AURORA2 BTAK PPP1R47 STK15 STK6 STK7 ARK-1 IPL1-related kinase aurora 2 aurora-related kinase 1 aurora/IPL1-like kinase aurora/IPL1-related kinase 1 breast tumor-amplified kinase breast-tumor-amplified kinase hARK1 protein phosphatase 1 regulatory subunit 47 serine/threonine kinase 6 serine/threonine protein kinase 15 serine/threonine-protein kinase 15 serine/threonine-protein kinase 6 serine/threonine-protein kinase aurora-A; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A
Gene Aliases: AIK; AIRK1; ARK-1; ARK1; AURA; AURKA; AYK1; BTAK; IAK1; PPP1R47; STK15; STK6; STK7
UniProt ID: (Human) O14965
Entrez Gene ID: (Human) 6790, (Rat) 261730
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.