Novus Biologicals
Manufacturer Code:NBP222118SS
Catalog # NBP222118SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F8 |
Immunogen | Purified recombinant fragment of human Aurora A expressed in E. coli. [Uniprot: O14965] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AIKAurA ARK-1 ARK1EC 2.7.11.1 AURA aurora kinase AhARK1 aurora/IPL1-like kinase Aurora/IPL1-related kinase 1 Aurora-related kinase 1 Breast tumor-amplified kinase breast-tumor-amplified kinase BTAKAURORA2 EC 2.7.11 IPL1-related kinase serine/threonine kinase 6 serine/threonine protein kinase 15 Serine/threonine-protein kinase 15 serine/threonine-protein kinase 6 Serine/threonine-protein kinase aurora-A STK15serine/threonine kinase 15 STK6MGC34538 STK7; ARK-1; Aurora 2; Aurora kinase A; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; Breast tumor-amplified kinase; protein phosphatase 1, regulatory subunit 47; serine/threonine protein kinase 15; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A
Gene Aliases: AIK; AIRK1; ARK1; AURA; AURKA; AYK1; BTAK; IAK1; PPP1R47; STK15; STK6; STK7
UniProt ID: (Human) O14965
Entrez Gene ID: (Human) 6790
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.