Proteintech
Manufacturer Code:102971AP150UL
Catalog # 50-172-6188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Aurora A Fusion Protein Ag0362 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIK ARK 1 ARK1 AURA AURKA Aurora A aurora kinase A Aurora related kinase 1 Aurora/IPL1 related kinase 1 AURORA2 Breast tumor amplified kinase BTAK hARK1 STK15 STK6 STK7; ARK-1; Aurora 2; aurora A; Aurora family kinase 1; Aurora kinase A; aurora-A; aurora-related kinase 1; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; Breast tumor-amplified kinase; Ipl1- and aurora-related kinase 1; protein phosphatase 1, regulatory subunit 47; ratAurA; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1
Gene Aliases: AIK; Airk; AIRK1; ARK-1; ARK1; AU019385; AURA; AURKA; Aurora-A; AW539821; AYK1; BTAK; IAK; IAK1; PPP1R47; STK15; STK6; STK7
UniProt ID: (Human) O14965, (Mouse) P97477
Entrez Gene ID: (Human) 6790, (Mouse) 20878, (Rat) 261730
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.