Novus Biologicals
Manufacturer Code:NBP221661
Catalog # NBP221661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of Drosophila melanogaster Aubergine/Sting protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aub; Aub aubergine CG6137 DmelCG6137 ms(2)32D sti sting; aub-PA; aub-PC; CG6137-PA; CG6137-PC; Protein aubergine; Protein sting; stellate-interacting gene
Gene Aliases: AUB; CG6137; Dmel\CG6137; Dmel_CG6137; ms(2)32D; sti; sting
UniProt ID: (Fruit fly) O76922
Entrez Gene ID: (Fruit fly) 34524
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.