Novus Biologicals
Manufacturer Code:NBP242817
Catalog # NBP242817
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (Na[+] + K[+])-ATPase alpha-subunit; (Na[+],K[+])ATPase alpha subunit; alpha-subunit of ATPase; ATPalpha; Atpalpha-PA; Atpalpha-PB; Atpalpha-PC; Atpalpha-PD; Atpalpha-PE; Atpalpha-PF; Atpalpha-PG; Atpalpha-PH; Atpalpha-PI; Atpalpha-PJ; Atpalpha-PK; ATPase alpha; ATPase alpha subunit; CG5670-PA; CG5670-PB; CG5670-PC; CG5670-PD; CG5670-PE; CG5670-PF; CG5670-PG; CG5670-PH; CG5670-PI; CG5670-PJ; CG5670-PK; Na(+)/K(+) ATPase alpha subunit; Na+,K+ ATPase alpha-subunit; Na, K ATPase a subunit; Na,K ATPase; Na,K ATPase alpha subunit; Na,K-ATPase; Na,K-ATPase alpha; Na,K-ATPase alpha subunit; Na,K-ATPase alpha-subunit; Na[ +], K[ +] -ATPase; Na[+] pump alpha subunit; Na[+], K[+] ATPase alpha; Na[+],K[+] ATPase alpha subunit; Na[+],K[+]-ATPase alpha subunit; Na[+]-K[+] ATPase; Na[+]/K[+] ATPase (alpha subunit); Na[+]/K[+] ATPase alpha subunit; Na[+]K[+] ATPase alpha subunit; Na[+]K[+]ATPase; NaK-ATPase; sodium pump alpha subunit; sodium pump alpha-subunit; Sodium pump subunit alpha; Sodium/potassium-transporting ATPase subunit alpha
Gene Aliases: (Na[+]/k[+])-ATPase; alpha-ATPase; Atp; Atp a; ATP-alpha; ATPa; ATPalpha; ATPBagr; CG5670; Da-47; Dmel\CG5670; Dmel_CG5670; dnATPase; JY; K ATPase; K-ATPase; K[+] -ATPase; K[+]-ATPase; K[+]-ATPase alpha subunit; K[+]-ATPase alpha-subunit; l(3)01164; l(3)01453; l(3)04492; l(3)j3B2; l(3)j3B7; l(3)j6A4; l(3)rM454; l(3)ry136; Na; Na+-K+-ATPase; Na+/K+ ATPase alpha subunit; Na+/K+-ATPase; Na-p; Na/K ATPase; Na/K-ATPase; Na/K-ATPase alpha-subunit; Na[+]; Na[+]-K[+]-ATPase alpha[[5]]-subunit; Na[+]/K[+] ATPase; Na[+]/K[+]-ATPase; Na[+]/K[+]ATPase; Na[+]K[+] ATPase; Na[+]K[+]-ATPase; Na[+]K[+]ATPase; NaK; NaK ATPase; NaK-ATPase; NKA; sodium/potassium transporting ATPase alpha
UniProt ID: (Fruit fly) P13607
Entrez Gene ID: (Fruit fly) 48971
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.