Novus Biologicals
Manufacturer Code:NBP276807
Catalog # NBP276807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SEREGHSINTR from the internal region of the protein sequence according to NP_002964.3 NP_001297050.1 NP_001297052.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: ataxin 2 ataxin 2) ataxin-2 ATX2spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2 autosomal dominant SCA2FLJ46772 Spinocerebellar ataxia type 2 protein TNRC13 trinucleotide repeat containing 13 Trinucleotide repeat-containing gene 13 protein; Ataxin-2; Spinocerebellar ataxia type 2 protein; Trinucleotide repeat-containing gene 13 protein
Gene Aliases: ASL13; ATX2; ATXN2; SCA2; TNRC13
UniProt ID: (Human) Q99700
Entrez Gene ID: (Human) 6311
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.