Novus Biologicals
Manufacturer Code:NB10058797
Catalog # NB10058797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1262 and 1312 of human ataxin 2 (spinocerebellar ataxia 2) using the numbering given in entry NP_002964.2 (GeneID 6311). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ataxin 2 ataxin 2) ataxin-2 ATX2spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2 autosomal dominant SCA2FLJ46772 Spinocerebellar ataxia type 2 protein TNRC13 trinucleotide repeat containing 13 Trinucleotide repeat-containing gene 13 protein; Ataxin-2; Spinocerebellar ataxia type 2 protein; Trinucleotide repeat-containing gene 13 protein
Gene Aliases: ASL13; ATX2; ATXN2; SCA2; TNRC13
UniProt ID: (Human) Q99700
Entrez Gene ID: (Human) 6311
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.