Invitrogen
Manufacturer Code:MA527667
Catalog # PIMA527667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S65-37 |
Immunogen | Synthetic peptide amino acids 746-761 (RKRRWSAPETRKLEKS) of mouse ataxin-1. Rat: 93&percnt identity (15/16 amino acids identical). Human: 87&percnt identity (14/16 amino acids identical). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alternative ataxin1; Ataxin-1; Ataxin-1 ATX1 Atxn1 D6S504E OTTHUMP00000016065 SCA1 Spinocerebellar ataxia type 1 protein Spinocerebellar ataxia type 1 protein homolog; spinocerebellar ataxia 1; spinocerebellar ataxia 1 homolog; Spinocerebellar ataxia type 1 protein; Spinocerebellar ataxia type 1 protein homolog
Gene Aliases: 2900016G23Rik; ATX1; ATXN1; C85907; D6S504E; ENSMUSG00000074917; Gm10786; SCA1
UniProt ID: (Human) P54253, (Mouse) P54254, (Rat) Q63540
Entrez Gene ID: (Human) 6310, (Mouse) 20238, (Rat) 25049
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.