Invitrogen
Manufacturer Code:PA578416
Catalog # PIPA578416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of mouse Ataxin 1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900016G23Rik Atx1 Atxn1 C85907 ENSMUSG00000074917 Gm10786 Sca1; Ataxin-1; spinocerebellar ataxia 1 homolog; Spinocerebellar ataxia type 1 protein homolog
Gene Aliases: 2900016G23Rik; Atx1; Atxn1; C85907; ENSMUSG00000074917; Gm10786; Sca1
UniProt ID: (Mouse) P54254
Entrez Gene ID: (Mouse) 20238
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.