Novus Biologicals
Manufacturer Code:NBP288942
Catalog # NB036537
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL10078 |
Immunogen | This antibody was developed against a synthetic peptide corresponding to amino acids: EEREMRFPTHPKESETEDQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Astrin; Astrin Deepest hMAP126 MAP126mitotic spindle coiled-coil related protein mitotic spindle associated protein p126 Mitotic spindle-associated protein p126 sperm associated antigen 5 sperm-associated antigen 5; Deepest; MAP126; mitotic spindle associated protein p126; mitotic spindle coiled-coil related protein; Mitotic spindle-associated protein p126; Sperm-associated antigen 5
Gene Aliases: DEEPEST; hMAP126; MAP126; SPAG5
UniProt ID: (Human) Q96R06
Entrez Gene ID: (Human) 10615
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.