Invitrogen
Manufacturer Code:MA526717
Catalog # PIMA526717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 77-313 of ASPA produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACY-2; ACY2 ASP ACY-2 aminoacylase 2 aminoacylase-2; aminoacylase 2; Aminoacylase-2; Aspartoacylase
Gene Aliases: ACY2; ASP; ASPA
UniProt ID: (Human) P45381
Entrez Gene ID: (Human) 443
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.