Novus Biologicals
Manufacturer Code:NBP270215G
Catalog # NBP270215G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A10 |
Immunogen | Full length recombinant protein of human Asparagine synthetase (NP_597680) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Asparagine synthetase [glutamine-hydrolyzing]; asparagine synthetase asparagine synthetase (glutamine-hydrolyzing) asparagine synthetase [glutamine-hydrolyzing] Cell cycle control protein TS11 EC 6.3.5.4 Glutamine-dependent asparagine synthetase TS11 TS11 cell cycle control protein; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11 cell cycle control protein
Gene Aliases: ASNS; ASNSD; TS11
UniProt ID: (Human) P08243
Entrez Gene ID: (Human) 440
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.