Novus Biologicals
Manufacturer Code:NBP187444
Catalog # NBP187444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QHFEFEYQTKVDGEIILHLYDKGGIEQTICMLDGVFAFVLLDTANKKVFLGRDTYGVRPLFKAMTEDGFLAVCSEAKGLVTLKHSATPFLKVEPFLPGHYEVLDLKPNGKVASVEMVKYHHCRDVP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Asparagine synthetase [glutamine-hydrolyzing]; asparagine synthetase asparagine synthetase (glutamine-hydrolyzing) asparagine synthetase [glutamine-hydrolyzing] Cell cycle control protein TS11 EC 6.3.5.4 Glutamine-dependent asparagine synthetase TS11 TS11 cell cycle control protein; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11 cell cycle control protein
Gene Aliases: ASNS; ASNSD; TS11
UniProt ID: (Human) P08243
Entrez Gene ID: (Human) 440
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.