Abnova
Manufacturer Code:MAB5646
Catalog # 89-112-341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8D7 |
Immunogen | Native purified rat liver membrane extracts Asgr1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse monoclonal antibody raised against native Asgr1.
For Research Use Only
Protein Aliases: ASGP-R 1; ASGPR 1; ASGR; asialoglycoprotein receptor (RHL1); Asialoglycoprotein receptor 1; Asialoglycoprotein receptor 1 (hepatic lectin); asialoglycoprotein receptor RHL1; Hepatic lectin 1; hepatic lectin 1 RHL1; hepatic lectin 1, RHL1; HL-1; MGC108731; RATRHL1; RHL-1; RHL1
Gene Aliases: ASGR; Asgr-1; Asgr1; RATRHL1; RHL1
UniProt ID: (Rat) P02706
Entrez Gene ID: (Rat) 24210
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.