Novus Biologicals
Manufacturer Code:NBP121701H
Catalog # NBP121701H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0074-6G32 |
Immunogen | Mouse recombinant Artemin |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: Artemin; artemin ENOVIN EVNneurotrophic factor NBN Neublastin; Enovin; Neublastin; neurotrophic factor
Gene Aliases: ARTN; ENOVIN; EVN; NBN
UniProt ID: (Human) O95441
Entrez Gene ID: (Human) 9048
Molecular Function:
growth factor
neurotrophic factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.