Invitrogen
Manufacturer Code:PA576306
Catalog # PIPA576306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide of human Artemin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Artemin; Enovin; EVN NBN ENOVIN; Neublastin; neurotrophic factor
Gene Aliases: ARTN; ENOVIN; EVN; NBN; neublastin
UniProt ID: (Human) O95441, (Rat) Q6AYE8, (Mouse) Q9Z0L2
Entrez Gene ID: (Human) 9048, (Rat) 362572, (Mouse) 11876
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.