Novus Biologicals
Manufacturer Code:NB300161SS
Catalog # NB018142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Amino acids GPGSRARAAGARGC of human Artemin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Artemin; artemin ENOVIN EVNneurotrophic factor NBN Neublastin; Enovin; Neublastin; neurotrophic factor
Gene Aliases: ARTN; ENOVIN; EVN; NBN
UniProt ID: (Human) O95441
Entrez Gene ID: (Human) 9048
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.