Proteintech
Manufacturer Code:215051AP150UL
Catalog # 50-173-3453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Arp5/ACTR5 Fusion Protein Ag15976 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin related protein 5 ACTR5 Arp5 hARP5 Sarcoma antigen NY SAR 16; Actin-related protein 5; ARP5 actin-related protein 5 homolog; hARP5; INO80 complex subunit M; Sarcoma antigen NY-SAR-16
Gene Aliases: AA545173; ACTR5; ARP5; B430109J19Rik; INO80M
UniProt ID: (Human) Q9H9F9
Entrez Gene ID: (Human) 79913, (Mouse) 109275, (Rat) 362258
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.