Novus Biologicals
Manufacturer Code:NBP261939R
Catalog # NBP261939R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H4 |
Immunogen | Synthetic peptide corresponding to AAs 376-390 of human aromatase (KALEDDVIGYPVKK) |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ARO1EC 1.14.14.1 Aromatase AROsubfamily XIX (aromatization of androgens) CYARMGC104309 CYPXIX cytochrome P450 family 19 subfamily A polypeptide 1 Cytochrome P-450AROM Estrogen synthase microsomal monooxygenase; Aromatase; CYPXIX; Cytochrome P-450AROM; Cytochrome P450 19A1; cytochrome P450, family 19, subfamily A, polypeptide 1; cytochrome P450, subfamily XIX (aromatization of androgens); Estrogen synthase; estrogen synthetase; flavoprotein-linked monooxygenase; microsomal monooxygenase
Gene Aliases: ARO; ARO1; CPV1; CYAR; CYP19; CYP19A1; CYPXIX; P-450AROM
UniProt ID: (Human) P11511
Entrez Gene ID: (Human) 1588
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.