Invitrogen
Manufacturer Code:PA586466
Catalog # PIPA586466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 211-260 of Human CYP19A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aromatase; Aromatase CYPXIX Cytochrome P-450AROM Cytochrome P450 19A1 Estrogen synthase CYP19A1 ARO1 CYAR CYP19; CYPXIX; Cytochrome P-450AROM; Cytochrome P450 19A1; cytochrome P450 family 19 subfamily a; Cytochrome P450, 19, aromatase; cytochrome P450, family 19, subfamily A, polypeptide 1; cytochrome P450, subfamily 19; cytochrome P450, subfamily XIX (aromatization of androgens); Estrogen synthase; estrogen synthetase; flavoprotein-linked monooxygenase; microsomal monooxygenase; P-450AROM
Gene Aliases: Ar; ArKO; ARO; ARO1; Arom; Aromatase; CPV1; CYAR; CYP19; Cyp19a; CYP19A1; CYPXIX; Int-5; Int5; P-450AROM; p450arom
UniProt ID: (Human) P11511, (Mouse) P28649
Entrez Gene ID: (Human) 1588, (Mouse) 13075, (Rat) 25147
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.