Invitrogen
Manufacturer Code:PA579125
Catalog # PIPA579125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of rat AromatasE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aromatase; CYPXIX; cytochrome P-450AROM; cytochrome P450 19A1; cytochrome P450 family 19 subfamily a; Cytochrome P450, 19, aromatase; cytochrome P450, subfamily 19; estrogen synthase; estrogen synthetase; P-450AROM
Gene Aliases: Aromatase; Cyp19; Cyp19a; p450arom
Entrez Gene ID: (Rat) 25147
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.