Invitrogen
Manufacturer Code:PA567021
Catalog # PIPA567021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Argonaute 4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: argonaute 4; Argonaute RISC catalytic component 4; Argonaute4; eIF-2C 4; eIF2C 4; Eukaryotic translation initiation factor 2C 4; Eukaryotic translation initiation factor 2C 4 AGO4 EIF2C4; eukaryotic translation initiation factor 2C, 4; hAgo4; Protein argonaute-4
Gene Aliases: AGO4; EIF2C4; KIAA1567
UniProt ID: (Human) Q9HCK5
Entrez Gene ID: (Human) 192670
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.